Calcein Orange™ sodium salt
Proveedor: AAT BIOQUEST
22008
AATB22008EA 307 EUR
AATB22008
Calcein Orange™ sodium salt
Calcein Orange™ sodium salt
Calcein AM is one of the most popular fluorescent probes used for labelling and monitoring cellular functions of live cells.
  • Low cytotoxicity
  • Improved photostability
  • Brighter fluorescence
  • Suitable for multiplexing
Precaución: For research use only.