CytoCalcein™ Violet 450 maximum excitation at 405 nm
Proveedor: AAT BIOQUEST
22012
AATB22012EA 307 EUR
AATB22012
CytoCalcein™ Violet 450 maximum excitation at 405 nm
CytoCalcein™ Violet 450
CytoCalcein™ Violet 450 is designed for labelling live cells in the same way to calcein, AM.
  • Optimized for flow cytometry excitation
  • Brighter signal
  • Suitable for multiplexing
Precaución: For research use only.