Tetramethylrhodamine methyl ester perchlorate (TMRM) ≥95%

Proveedor: AAT BIOQUEST
Atención

Sinónimos: N-[6-(Dimethylamino)-9-[2-(methoxycarbonyl)phenyl]-3H-xanthen-3-ylidene]-N-methylmethanaminium Perchlorate, Tetramethylrhodamine methyll ester Perchlorate, 3,6-Bis(dimethylamino)-9-(2-(methoxycarbonyl)phenyl)xanthylium perchlorate, Tetramethylrhodamine Methyl Ester Perchlorate

22221
AATB22221EA 149 EUR
AATB22221
Tetramethylrhodamine methyl ester perchlorate (TMRM) ≥95%
Tetramethylrhodamine methyl ester perchlorate (TMRM)

Positively charged rhodamine dyes (such as rhodamine esters and rosamines) are selectively localized in mitochondria, thus they are widely used for labeling mitochondria of live cells.

Formula: C₂₅H₂₅N₂O₃·ClO₄
MW: 500.93 g/mol
Número MDL: MFCD00274460
Núm. CAS: 115532-50-8
ADR: 5 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR