Screen Quest™ Fluo-8 Medium Removal Calcium Assay Kit, Optimized for Difficult Cell Lines
Proveedor: AAT BIOQUEST
36307 36308 36309
AATB36307EA 152 EUR
AATB36307 AATB36308 AATB36309
Screen Quest™ Fluo-8 Medium Removal Calcium Assay Kit, Optimized for Difficult Cell Lines
Ensayos Ensayos celulares
Calcium flux assays are preferred methods in drug discovery for screening G protein coupled receptors (GPCR).
  • 100 to 250 times fluorescence increase once bound to calcium
  • Contains all necessary components
  • Suitable for multiple types of cells
  • Robust and adaptable protocol
  • Brighter and more photostable signal
Precaución: For research use only.