Native Horseradish Horseradish peroxidase (from Serum), Ofloxacin
Proveedor: AAT BIOQUEST
50019
AATB50019EA 769 EUR
AATB50019
Native Horseradish Horseradish peroxidase (from Serum), Ofloxacin
Proteínas y Péptidos
Ofloxacin-HRP conjugate is used for developing an ELISA assay for quantifying ofloxacin.
  • High immunogenicity
  • Useful for antibody development
  • ELISA assay building block
Precaución: For research use only.