Native Bovine Bovine Serum Albumin (from Serum), Florfenicol
Proveedor: AAT BIOQUEST
50035
AATB50035EA 769 EUR
AATB50035
Native Bovine Bovine Serum Albumin (from Serum), Florfenicol
Proteínas y Péptidos
Florfenicol-BSA conjugate is used for developing anti-florfenicol antibodies that are typically used in an ELISA assay.
  • High immunogenicity
  • Useful for antibody development
  • ELISA assay building block
Precaución: For research use only.