Anti-IgG Goat Polyclonal Antibody

Proveedor: AAT BIOQUEST
50269
AATB50269EA 115 EUR
AATB50269
Anti-IgG Goat Polyclonal Antibody
Anticuerpos
AAT Bioquest's anti-human secondary antibodies have well-characterised specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target.

  • Purified and quality-tested for consistent results
  • Suitable for multiple applications
  • Purified through affinity chromatography

Multiple secondary antibodies can bind to a single primary antibody.

Precaución: Research use only.

Type: Control
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: IgG
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR