ReadiLink™ Rapid XFD594 Antibody Labelling Kit
Proveedor: AAT BIOQUEST
5736
AATB5736EA 687 EUR
AATB5736
ReadiLink™ Rapid XFD594 Antibody Labelling Kit
Sistemas de enlace y etiquetado de proteínas
ReadiLink™ Rapid antibody labelling kits, designed for production scale, provide a convenient and efficient method for labelling large volumes of antibodies with XFD dyes (equivalent to Alexa Fluor®).
  • Robust protocol
  • Consistent results
  • Six-month shelf life
Designed to provide high quality material for downstream fluorescent applications, including flow cytometry.
Información de suministro: Shipped at RT.
Precaución: Research use only.