JUN Dye qPCR Calibration Solution *10000X*
Proveedor: AAT BIOQUEST
67038
AATB67038EA 420 EUR
AATB67038
JUN Dye qPCR Calibration Solution *10000X*
Reactivos de ácidos nucleicos qPCR/RT-qPCR Enzymes and Kits
JUN dye qPCR Calibration Solution supplied as a 10000X concentrate is compatible with a wide variety of qPCR instruments, including the ABI7500 Fast, QuantStudio™, and ViiA™ 7 systems.
  • Stable for 12 months
Real-time thermal cyclers require calibration to generate a spectral profile for the fluorescence signal.
Precaución: For research use only.