Anti-ITPR1 Rabbit Polyclonal Antibody

Proveedor: Abcam
AB111615-100
ABCAAB111615-100EA 688 EUR
ABCAAB111615-100
Anti-ITPR1 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: ITPR1 (inositol 1,4,5-trisphosphate receptor, type 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR