Anti-Penicillin Mouse Monoclonal Antibody [clone: P2B9]

Proveedor: Abcam
AB116641-100
ABCAAB116641-100EA 530 EUR
ABCAAB116641-100
Anti-Penicillin Mouse Monoclonal Antibody [clone: P2B9]
Anticuerpos
Type: Primary
Antigen: Penicillin
Clonality: monoclonal
Clone: P2B9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR