Anti-PRKCG Rabbit Polyclonal Antibody

Proveedor: Abcam
AB4144-100
ABCAAB4144-100EA 914 EUR
ABCAAB4144-100
Anti-PRKCG Rabbit Polyclonal Antibody
Anticuerpos
Precaución:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production

More Product Information
Order Now

ESPECIFICACIONES

Simbolo antígeno PRKCG
Environmentally Preferable
Nombre del antígeno Protein Kinase C Gamma
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Unconjugated
Reactividad Human
Huésped Rabbit
Isotipo IgG
Western blot Yes
ELISA Yes
InmunoQuímica Yes
FluorescenciaInmunológica Yes
Temperatura de almacenamiento Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR