Anti-HDAC11 Rabbit Polyclonal Antibody

Proveedor: Abgent
AP1111A
ABGEAP1111AEA 354 EUR
ABGEAP1111A
Anti-HDAC11 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: HDAC11 (histone deacetylase 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

ESPECIFICACIONES

Simbolo antígeno HDAC11
Environmentally Preferable
Nombre del antígeno Histone deacetylase 11
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Unconjugated
Reactividad Human,Mouse
Huésped Rabbit
Isotipo
Western blot Yes
ELISA Yes
InmunoQuímica Yes
Epítopo N-Terminal (1–100 aa)
Adsorción cruzada No
Temperatura de almacenamiento Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR