Anti-LEF1 Rabbit Polyclonal Antibody

Proveedor: Abgent
AP12048A
ABGEAP12048AEA 354 EUR
ABGEAP12048A
Anti-LEF1 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: LEF1 (lymphoid enhancer-binding factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR