Anti-ASPG Rabbit Polyclonal Antibody

Proveedor: Abgent
AP13630C
ABGEAP13630CEA 354 EUR
ABGEAP13630C
Anti-ASPG Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: ASPG (asparaginase homolog (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

ESPECIFICACIONES

Simbolo antígeno ASPG
Environmentally Preferable
Nombre del antígeno Asparaginase
Sinónimos de antígenos ASPG
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Unconjugated
Reactividad Human
Huésped Rabbit
Isotipo
Western blot Yes
ELISA Yes
Epítopo middle
Temperatura de almacenamiento Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR