Anti-CSRNP3 Rabbit Polyclonal Antibody

Proveedor: Abgent
AP16212A
ABGEAP16212AEA 354 EUR
ABGEAP16212A
Anti-CSRNP3 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: CSRNP3 (cysteine-serine-rich nuclear protein 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR