Anti-MTMR6 Rabbit Polyclonal Antibody

Proveedor: Abgent
AP16543A
ABGEAP16543AEA 354 EUR
ABGEAP16543A
Anti-MTMR6 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: MTMR6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

ESPECIFICACIONES

Simbolo antígeno MTMR6
Nombre del antígeno Myotubularin Related Protein 6
Environmentally Preferable
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Unconjugated
Reactividad Human
Huésped Rabbit
ID del gen 9107
Isotipo
Western blot Yes
ELISA Yes
FluorescenciaInmunológica Yes
Epítopo N-terminal
Inmunógeno This MTMR6 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 114-143 amino acids from the N-terminal region of human MTMR6.
Número de aminoácidos 114 to 143
Sequence DLAEEYKRMGVPNSHC
Molecular weight 71.968 kDa
Purification Protein A & Peptide affinity purification
Concentración 0.7
Storage buffer pH 7,4
Temperatura de almacenamiento Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
Shipping temperature 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR