Anti-ARL6IP1 Rabbit Polyclonal Antibody

Proveedor: Abgent
AP16637A
ABGEAP16637AEA 354 EUR
ABGEAP16637A
Anti-ARL6IP1 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: ARL6IP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR