Anti-SLC2A9 Rabbit Polyclonal Antibody

Proveedor: Abgent
AP16871A
ABGEAP16871AEA 354 EUR
ABGEAP16871A
Anti-SLC2A9 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: SLC2A9 (solute carrier family 2 (facilitated glucose transporter), member 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR