Anti-SNCB Rabbit Polyclonal Antibody

Proveedor: Abgent
AP5842B
ABGEAP5842BEA 354 EUR
ABGEAP5842B
Anti-SNCB Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: SNCB (synuclein, beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR