Anti-Quinidine Monoclonal Antibody [clone: 11-265.2.3]

Proveedor: Abnova
MAB2249
ABNOMAB2249EA 402 EUR
ABNOMAB2249
Anti-Quinidine Monoclonal Antibody [clone: 11-265.2.3]
Anticuerpos
Mouse monoclonal antibody raised against Quinidine.

Type: Primary
Antigen: Quinidine
Clonality: Monoclonal
Clone: 11-265.2.3
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1
Reactivity:
Order Now

ESPECIFICACIONES

Environmentally Preferable
Nombre del antígeno Quinidine
Tipo de anticuerpo Primario
Clonalidad Monoclonal
Conjugado Unconjugated
Clon 11-265.2.3
Reactividad
Huésped
Isotipo IgG1
ELISA Yes
Inmunógeno Quinidine conjugated with KLH.
Formato Liquid
Adsorción cruzada No
Storage buffer PBS, pH 7,4
Temperatura de almacenamiento Store at – 80 °C. Aliquot to avoid repeated freezing and thawing.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR