Anti-BIRC5 Rabbit Polyclonal Antibody (DyLight® 549)

Proveedor: Abnova
PAB15180
ABNOPAB15180EA 779 EUR
ABNOPAB15180
Anti-BIRC5 Rabbit Polyclonal Antibody (DyLight® 549)
Anticuerpos
Rabbit polyclonal antibody raised against synthetic phosphopeptide of BIRC5.

Recommended Dilutions: Western Blot: 3 ?g/ml

Type:
Antigen: BIRC5
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR