Anti-SLIRP Rabbit Polyclonal Antibody (DyLight® 549)

Proveedor: Abnova
PAB15212
ABNOPAB15212EA 779 EUR
ABNOPAB15212
Anti-SLIRP Rabbit Polyclonal Antibody (DyLight® 549)
Anticuerpos
Rabbit polyclonal antibody raised against synthetic peptide of SLIRP.

Recommended Dilutions: Western Blot: 0.5-2 ?g/ml

Type:
Antigen: SLIRP
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Primates
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR