Anti-Florfenicol(N) Polyclonal Antibody

Proveedor: Abnova
PAB8275
ABNOPAB8275EA 1170 EUR
ABNOPAB8275
Anti-Florfenicol(N) Polyclonal Antibody
Anticuerpos
Sheep polyclonal antibody raised against Florfenicol(N).

Recommended Dilutions: Enzyme Immunoassay (2.5 ?g/ml

Type: Primary
Antigen: Florfenicol(N)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR