L(+)-Isoleucina 99%

Proveedor: Thermo Fisher Scientific

Sinónimos: Ile, I, (2S,3S)-(+)-Isoleucina

166170250 166171000 166170050 166175000
ACRO166170250EA 82 EUR
ACRO166170250 ACRO166171000 ACRO166170050 ACRO166175000
L(+)-Isoleucina 99%
L(+)-Isoleucina
Formula: H₃CCH₂CH(CH₃)CH(NH₂)COOH
MW: 131.17 g/mol
Pto. de fusión: 284…286 °C (desc.)
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00064222
Núm. CAS: 73-32-5
EINECS: 200-798-2
Índice Merck: 14,05179

Order Now

Resultados test especificaciones

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Loss on drying ≤0.3 % (105°C, 3 hrs)
Residue after ignition ≤0.3 %
Specific optical rotation +38.9° to +41.8° (20°C, 589 nm) (c=4, 6 N HCl)
Titration with HClO4 98.5 to 101.5 %
Chloride (Cl) ≤500 ppm
Sulfate (SO4) ≤300 ppm
Iron (Fe) ≤30 ppm
Heavy metals (as Pb) ≤20 ppm

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR