Phenylpyruvic acid sodium salt monohydrate 98%

Proveedor: Thermo Fisher Scientific

189730050 189730250
ACRO189730050EA 57 EUR
ACRO189730050 ACRO189730250
Phenylpyruvic acid sodium salt monohydrate 98%
Phenylpyruvic acid sodium salt monohydrate

Appearance: White to light yellow Powder

Formula: C₉H₇NaO₃·H₂O
Pto. de fusión: 260 °C
Temperatura de almacenaje: Nevera
Número MDL: MFCD00149109
Núm. CAS: 122049-54-1

Order Now

Resultados test especificaciones

Appearance White to light yellow Powder
Infrared spectrum Conforms
Water 7 to 12 % (K.F.)
Titration with HClO4 97.5 to 102.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR