Carmín alum lake of carminic acid, pure, high purity tinción biológica

Proveedor: Thermo Fisher Scientific

190200050 190200250 190201000
ACRO190200050EA 78.3 EUR
ACRO190200050 ACRO190200250 ACRO190201000
Carmín alum lake of carminic acid, pure, high purity tinción biológica
Carmín
Formula: C₂₂H₂₀O₁₃
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00167028
Núm. CAS: 1390-65-4
EINECS: 215-724-4

Order Now

Resultados test especificaciones

Appearance Dark red to purple Powder
Infrared spectrum Conforms
UV spectrum Complies
Assay ≥42.0 % (Carminic acid)
Loss on drying ≤20 %
Ash ≤12 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR