Xanthosine dihydrate 99%

Proveedor: Thermo Fisher Scientific

Sinónimos: 9-β-D-Ribofuranosylxanthine dihydrate, 9-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-9H-purine-2,6-diol dihydrate, 3,9-Dihydro-9-beta-D-ribofuranosyl-1H-purine-2,6-dione dihydrate

207630010 207630050
ACRO207630010EA 57 EUR
ACRO207630010 ACRO207630050
Xanthosine dihydrate 99%
Xanthosine dihydrate
Formula: C₁₀H₁₂N₄O₆·2H₂O
Temperatura de almacenaje: Nevera
Número MDL: MFCD00149347
Núm. CAS: 5968-90-1

Order Now

Resultados test especificaciones

Appearance White to off-white Powder
Infrared spectrum Conforms
Water 9 to 14 %
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR