3-Buten-1-ol 98+%

Proveedor: Thermo Fisher Scientific

295750250 295751000 295755000
ACRO295750250EA 142 EUR
ACRO295750250 ACRO295751000 ACRO295755000
3-Buten-1-ol 98+%
3-Buten-1-ol
Formula: C₄H₈O
Número MDL: MFCD00002959
Núm. CAS: 627-27-0

Order Now

Resultados test especificaciones

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4200 to 1.4220 (20°C, 589 nm)
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR