(Triphenylphosphoranylidene)acetaldehyde 97%

Proveedor: Thermo Fisher Scientific
Atención

Sinónimos: 2-(Triphenylphosphoranylidene)acetaldehyde, (Formylmethylene)triphenylphosphorane

307490050 307490250 307491000
ACRO307490050EA 69.2 EUR
ACRO307490050 ACRO307490250 ACRO307491000
(Triphenylphosphoranylidene)acetaldehyde 97%
(Triphenylphosphoranylidene)acetaldehyde

CAS No.: 2136-75-6

Formula: C₂₀H₁₇OP
MW: 304.33 g/mol
Pto. de fusión: 185…188 °C
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00006994
Núm. CAS: 2136-75-6

Order Now

Resultados test especificaciones

Appearance Cream to pink to orange to brown Crystalline powder
Infrared spectrum Conforms
Water ≤4.0 % (K.F.)
Titration with HClO4 96.0 to 104.0 % (on dry substance)

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR