Nω-Nitro-L-arginine methyl ester hydrochloride 98%

Proveedor: Thermo Fisher Scientific

Sinónimos: N-ω-Nitro-L-Arginine Methyl Ester Hydrochloride, L-NAME Hydrochloride, L-NAME HCl, H-Arg(NO2)-OMe.HCl, (S)-Methyl 2-amino-5-(3-nitroguanidino)pentanoate hydrochloride

328750050 328750250
ACRO328750050EA 144 EUR
ACRO328750050 ACRO328750250
Nω-Nitro-L-arginine methyl ester hydrochloride 98%
Nomega-Nitro-L-arginine methyl ester hydrochloride

CAS No.: 51298-62-5

Formula: C₇H₁₅N₅O₄
Pto. de fusión: 159…161 °C
Temperatura de almacenaje: Nevera
Número MDL: MFCD00039052
Núm. CAS: 51298-62-5

Order Now

Resultados test especificaciones

Appearance White to off-white Crystalline solid
Infrared spectrum Conforms
HPLC ≥97.5 %
Specific optical rotation +14.0° to +16.0° (24°C, 589 nm) (c=3, CH3OH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR