Creatina fosfato tetrahidrato 99%

Proveedor: Thermo Fisher Scientific

337340100 337340010 337340020
ACRO337340100EA 296 EUR
ACRO337340100 ACRO337340010 ACRO337340020
Creatina fosfato tetrahidrato 99%
Fosfocreatina sal sodica tetrahidrato
Formula: C₄H₈N₃Na₂O₅P·4H₂O
Temperatura de almacenaje: Nevera
Número MDL: MFCD00150192
Núm. CAS: 71519-72-7

Order Now

Resultados test especificaciones

Appearance White Powder
Infrared spectrum Conforms
Water 21 to 25 % (K.F.)
Solubility (10 % in water) Clear colorless solution
Titration with HClO4 98.5 to 101.5 %
Heavy metals (as Pb) ≤20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR