Ruthenium(III) nitrosylnitrate in diluted aq. nitric acid solution

Proveedor: Thermo Fisher Scientific
Peligro

365040250 365041000 365045000
ACRO365040250EA 94.9 EUR
ACRO365040250 ACRO365041000 ACRO365045000
Ruthenium(III) nitrosylnitrate in diluted aq. nitric acid solution
Ruthenium(III) nitrosylnitrate

Appearance: Clear orange to dark brown Solution

Formula: N₄O₁₀Ru
Densidad: 1,07 g/cm³
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00016313
Núm. CAS: 34513-98-9
UN: 3093
ADR: 8,II

Order Now

Resultados test especificaciones

Appearance Clear orange to dark brown Solution
Assay 1.5 % Ru (typically)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR