α-Cloro-3,4-difluorotolueno 98%

Proveedor: Thermo Fisher Scientific
Peligro

384420050 384420250
ACRO384420050EA 82.6 EUR
ACRO384420050 ACRO384420250
α-Cloro-3,4-difluorotolueno 98%
α-Cloro-3,4-difluorotolueno
Formula: C₇H₅ClF₂
Número MDL: MFCD00236218
Núm. CAS: 698-80-6
UN: 3261
ADR: 8,II

Order Now

Resultados test especificaciones

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4920 to 1.4940 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR