Trimethylphenylammonium hydroxide 0.1 M in methanol

Proveedor: Thermo Fisher Scientific
Peligro

422021000 422025000 422020025
ACRO422021000EA 148 EUR
ACRO422021000 ACRO422025000 ACRO422020025
Trimethylphenylammonium hydroxide 0.1 M in methanol
Trimethylphenylammonium hydroxide

CAS No.: 1899-02-1

Formula: C₉H₁₅NO
Densidad: 0,8 g/cm³
Pto. de inflamación: 10 °C
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00041899
Núm. CAS: 1899-02-1
UN: 1230
ADR: 3,II

Order Now

Resultados test especificaciones

Appearance Clear colorless to light yellow Solution
Strength 0.09 to 0.11 M
Melting point 69 to 73 °C
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR