2-Methoxy-3-pyridineboronic acid 97%

Proveedor: Thermo Fisher Scientific
Atención

430250010 430250050
ACRO430250010EA 51 EUR
ACRO430250010 ACRO430250050
2-Methoxy-3-pyridineboronic acid 97%
2-Methoxy-3-pyridineboronic acid
Formula: C₆H₈BNO₃
Pto. de fusión: 143…145 °C
Temperatura de almacenaje: Nevera
Número MDL: MFCD03411572
Núm. CAS: 163105-90-6

Order Now

Resultados test especificaciones

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 141 to 146 °C
HPLC ≥96.0 %
Residue after evaporation ≤0.0005 %
Acidity ≤0.0005 meq/g
Free chlorine ≤0.00003 % (as Cl)
Aluminium (Al) ≤0.5 ppm
Barium (Ba) ≤0.1 ppm
Cadmium (Cd) ≤0.05 ppm
Calcium (Ca) ≤0.5 ppm
Chromium (Cr) ≤0.02 ppm
Cobalt (Co) ≤0.02 ppm
Copper (Cu) ≤0.02 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.1 ppm
Magnesium (Mg) ≤0.1 ppm
Manganese (Mn) ≤0.02 ppm
Nickel (Ni) ≤0.02 ppm
Tin (Sn) ≤0.1 ppm
Zinc (Zn) ≤0.1 ppm
Aldehyde (HCHO) ≤0.0005 %
Ethanol ≤0.3 % (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR