Ciprofloxacin hydrochloride hydrate 98%

Proveedor: Thermo Fisher Scientific

456880050 456880250
ACRO456880050EA 40.3 EUR
ACRO456880050 ACRO456880250
Ciprofloxacin hydrochloride hydrate 98%
Ciprofloxacin hydrochloride hydrate
Formula: C₁₇H₁₈FN₃O₃·HCl·xH₂O
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00242856
Núm. CAS: 128074-72-6

Order Now

Resultados test especificaciones

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Water 4.0 to 7.0 %
HPLC ≥97.5 %
Residue after ignition ≤0.1 %

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR