Vitamin B12a hydrochloride

Proveedor: Thermo Fisher Scientific

Sinónimos: Hydroxocobalamin hydrochloride, Hydroxocobalamin xhydrochloride

462080010 462080050
ACRO462080010EA 337 EUR
ACRO462080010 ACRO462080050
Vitamin B12a hydrochloride
Vitamin B12a hydrochloride

  • Bioactive

Formula: C₆₂H₈₉CoN₁₃O₁₅P·HCl
MW: 1346.35 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00167463
Núm. CAS: 58288-50-9

Order Now

Resultados test especificaciones

Appearance Dark red to dark purple Crystalline powder or crystals
Infrared spectrum Conforms
Assay ≥85 %
Loss on drying ≤12 %
HPLC ≥90 %
Identification Chlorides: passes test
UV Lambda max 525 nm
Absorbance scale 0.31 to 0.35 (A525/A351)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR