Tylosin tartrate

Proveedor: Thermo Fisher Scientific
Peligro

Sinónimos: Pharmasin, [R-(R'R')]-2,3-Dihydroxybutanedionate Tylosin salt

463070050 463070250 463071000
ACRO463070050EA 119 EUR
ACRO463070050 ACRO463070250 ACRO463071000
Tylosin tartrate
Tylosin tartrate

  • Bioactive

Formula: C₄₆H₇₇NO₁₇·C₄H₆O₆
MW: 1066.20 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00070094
Núm. CAS: 74610-55-2

Order Now

Resultados test especificaciones

Appearance White to light yellow Powder
Infrared spectrum Conforms
Solubility (5 % in water) Clear light yellow to yellow-green
HPLC ≥94 % (sum of tylosins A+B+C+D)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR