Multi-element quality control standards for ICP
Proveedor: CPACHEM
M8A96.1.5N.L1 M8A96.1.5N.L5
ACSDM8A96.1.5N.L1EA 123 EUR
ACSDM8A96.1.5N.L1 ACSDM8A96.1.5N.L5
Multi-element quality control standards for ICP
Normativas Estándares catiónicos Estándares de multielementos
ICP QC standards used for Al, Ag, As, B, Ba, Be, Bi, Ca, Cd, Cs, Co, Cr, Cu, Fe, In, K, Li, Mg, Mn, Mo, Na, Ni, Nb, Pb, Rb, Sb, Se, Sr, Ti, Tl, V, U, Zn. 1 mg/l for 33 elememts.