Suction cannulae, Aesculap®
Proveedor: Aesculap
MD124R MD125R MD127R MD129R
AESCMD124REA 54.7 EUR
AESCMD124R AESCMD125R AESCMD127R AESCMD129R
Suction cannulae, Aesculap®
Cánulas
Suction cannulae with finger cut-off and stylet.
  • Author: Frazier
  • Thumb piece 90° to curve
  • With LUER hub