BJ5183 Electroporation-competent cells
Proveedor: STRATAGENE
200157 200154 200157
STRA200157EA 0 EUR
STRA200157 AGLS200154 AGLS200157
BJ5183 Electroporation-competent cells
Células competentes
BJ5183 electroporation competent cells are a recombination proficient bacterial strain.
  • 300% more recombinants with specialty competent cells
  • Recombination proficient bacterial cell strain
  • Infects dividing and non-dividing cells
These cells supply the components necessary to execute the recombination event between a transfer vector containing the gene of interest and a vector containing the adenoviral genome, provided that appropriate regions of homology are shared between the two vectors.
BJ5183 cells are usually cotransformed with a transferMore Product Information