XL1 blue and XL2 blue competent cells
Proveedor: STRATAGENE
200130 200151 200158 200130 200150 200151 200228 200229 200236 200249
STRA200130EA 0 EUR
STRA200130 STRA200151 STRA200158 AGLS200130 AGLS200150 AGLS200151 AGLS200228 AGLS200229 AGLS200236 AGLS200249
XL1 blue and XL2 blue competent cells
Células competentes
XL1 and XL2-Blue Competent Cells are versatile for routine cloning and allow blue-white colour screening for high efficiency cloning of methylated DNA.
  • Designed to provide a host for optimal propagation of both plasmid and lambda phage vectors
  • Available in a range of cloning efficiencies
  • Available as economical subcloning-grade Competent Cells for routine procedures when DNA is not limited
  • Available as supercompetent cells in a variety of strains at efficiencies greater than 1×10⁹ transformants/μg.
  • Transformation efficiency for XL1-Blue MR and XL-1 Blue