Bioanalyzer RNA Analysis
Proveedor: AGILENT
5067-1511 5067-1512 5067-1529
AGLS5067-1511EA 1150 EUR
AGLS5067-1511 AGLS5067-1512 AGLS5067-1529
Bioanalyzer RNA Analysis
Sistemas de electroforesis Electroforesis capilar
Reliable and reproducible RNA sample quality assessment.
  • Maximum flexibility is enabled with options to check total RNA, mRNA, or cRNA quality
  • Minimal sample consumption is required with only 5 ng of total RNA needed for an effective analysis
  • Easily determine the RNA integrity number (RIN) using in-built software that automatically assigns an integrity value to a total RNA sample
  • This tool was designed to provide scientists an objective measurement of degradation in RNA samples
  • RNase free consumables avoid sample degradation during analysis
  • Accurate