Oligo Pro II System Capillary Arrays
Proveedor: AGILENT
A2500-12755580 A2500-24755580 A2500-96755580
AGLSA2500-12755580EA 2277 EUR
AGLSA2500-12755580 AGLSA2500-24755580 AGLSA2500-96755580
Oligo Pro II System Capillary Arrays
Sistemas de electroforesis Electroforesis capilar
Three different capillary arrays for analysis on the Oligo Pro II system.
  • High resolution - optimised length allowing for n-1 resolution through 60 nt
  • Variable throughput - 12, 24, and 96 capillary arrays designed to meet individual throughput demands
  • Designed for the Oligo Pro II system - compatible with UV detection allowing for visualisation of unlabeled oligonucleotides
The Oligo Pro II system has 3 capillary arrays, the 12-, 24-, and 96-Capillary Array Long, for analysis with the Oligo Pro Reagents. All three have a separation or effective lengthMore Product Information