BAC Kits
Proveedor: AGILENT
FP-1003-0275 FP-1004-0275
AGLSFP-1003-0275EA 1660 EUR
AGLSFP-1003-0275 AGLSFP-1004-0275
BAC Kits
Sistemas de electroforesis Electroforesis capilar
Quality control of BAC digests and large DNA fragments using pulsed-field capillary electrophoresis.
  • Reliable sizing - Accurate and precise sizing of large DNA fragments from 75 bp to 48 kb or 75 bp to 165 kb
  • High resolution separations - Optimised pulsed-field method prevents peak broadening, allowing separation of similarly sized large DNA fragments
  • Flag analysis - ProSize data analysis software allows for automated flagging of DNA fragments decreasing analysis times
  • Low sample volume - Only 2 µl required, conserving sample for downstream analysis
  • Fast