Anti-IgG (H+L) Chicken Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))

Proveedor: Agrisera
AS101244
AGRIAS101244EA 253 EUR
AGRIAS101244
Anti-IgG (H+L) Chicken Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Anticuerpos
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope:
Host: Chicken
Isotype:
Reactivity: Hamster
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR