Standard hydrometers
Proveedor: Alla France
3500LC06020BQP 3500LC06520BQP 3500LC14020HQP 3500LC14520HQP 3500LC15020HQP 3500LC16020HQP 3500LC16520HQP 3500LC17020HQP 3500LC17520HQP 3500LC18020HQP 3500LC19020HQP 3500LC19520HQP 3500MC06020BQP 3500MC06520BQP 3500MC07020BQP 3500MC07520BQP 3500MC12520MQP 3500MC13020HQP 3500MC13020MQP 3500MC13520HQP 3500MC13520MQP 3500MC15520MQP 3100MC06020BQP 3100MC140/20HQ 3100MC150/20HQ 3100MC16020HQP 3100MC17020HQP 3100MC18020HQP 3100MC19020HQP 3500LC18520HQP 3500MC16020HQP 3500MC16520HQP 3500MC17020HQP 3500MC17520HQP 3500MC18020HQP 3500MC18520HQP 3500MC19020HQP 3500MC19520HQP 3100MC07020BQP 3100MC08020BQP 3100MC09020BQP 3100MC10020BQP 3100MC11020BQP 3100MC12020BQP 3100MC13020BQP
ALLA3500LC06020BQPEA 57 EUR
ALLA3500LC06020BQP ALLA3500LC06520BQP ALLA3500LC14020HQP ALLA3500LC14520HQP ALLA3500LC15020HQP ALLA3500LC16020HQP ALLA3500LC16520HQP ALLA3500LC17020HQP ALLA3500LC17520HQP ALLA3500LC18020HQP ALLA3500LC19020HQP ALLA3500LC19520HQP ALLA3500MC06020BQP ALLA3500MC06520BQP ALLA3500MC07020BQP ALLA3500MC07520BQP ALLA3500MC12520MQP ALLA3500MC13020HQP ALLA3500MC13020MQP ALLA3500MC13520HQP ALLA3500MC13520MQP ALLA3500MC15520MQP 614-1523 614-1525 614-1526 614-1527 614-1528 614-1529 614-1530 ALLA3500LC18520HQP ALLA3500MC16020HQP ALLA3500MC16520HQP ALLA3500MC17020HQP ALLA3500MC17520HQP ALLA3500MC18020HQP ALLA3500MC18520HQP ALLA3500MC19020HQP ALLA3500MC19520HQP ALLA3100MC07020BQP ALLA3100MC08020BQP ALLA3100MC09020BQP ALLA3100MC10020BQP ALLA3100MC11020BQP ALLA3100MC12020BQP ALLA3100MC13020BQP
Standard hydrometers
Hidrómetros
These standard hydrometers strictly adhere to the principles of manufacture and specified calibration.
  • Reading below meniscus
  • Calibration temperature: 20 °C
  • Reference mark on the stem shows any movement of the measurement scale
Certificaciones: NF, ISO, BS and DIN.
Información para pedidos: Include three calibration points (factory certificate or accredited laboratory certificate on request). The calibration points should be specified when ordering.
Información de suministro: Supplied with certificate of conformity to ISO 649 (correspondingMore Product Information