Anti-A20 Mouse Monoclonal Antibody [clone: 59A426]

Proveedor: Merck Millipore (Oncogene)
AM63-100
AM63-100EA 633 EUR
AM63-100
Anti-A20 Mouse Monoclonal Antibody [clone: 59A426]
Anticuerpos
Type: Primary
Antigen: A20
Clonality: monoclonal
Clone: 59A426
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

ESPECIFICACIONES

Simbolo antígeno A20
Environmentally Preferable
Tipo de anticuerpo Primario
Clonalidad Monoclonal
Conjugado Unconjugated
Clon 59A426
Reactividad Human
Huésped Mouse
Isotipo
Western blot Yes
Precipitación inmunológica Yes
Temperatura de almacenamiento Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR