Precision thermometers, NS 14,5 / 23
Proveedor: Amarell
D262014-CB D262016-CB D262234-FL D262236-FL D262238-FL D262242-EF D262330-FL D262332-FL D262334-FL D262338-EF D262350-FL D262352-FL D262354-FL D262358-EF D262370-FL D262374-FL D262378-EF D262390-FL D262394-FL D262398-EF D262410-FL D262414-FL D262418-EF D262430-FL D262434-FL D262438-EF D262450-FL D262454-FL D262458-EF D262490-FL D262494-FL D262498-EF
AMARD262014-CBEA 0 EUR
AMARD262014-CB AMARD262016-CB AMARD262234-FL AMARD262236-FL AMARD262238-FL AMARD262242-EF AMARD262330-FL AMARD262332-FL AMARD262334-FL AMARD262338-EF AMARD262350-FL AMARD262352-FL AMARD262354-FL AMARD262358-EF AMARD262370-FL AMARD262374-FL AMARD262378-EF AMARD262390-FL AMARD262394-FL AMARD262398-EF AMARD262410-FL AMARD262414-FL AMARD262418-EF AMARD262430-FL AMARD262434-FL AMARD262438-EF AMARD262450-FL AMARD262454-FL AMARD262458-EF AMARD262490-FL AMARD262494-FL AMARD262498-EF
Precision thermometers, NS 14,5 / 23
Termómetros Analogue Thermometers
These precision thermometers with standard ground joint are suitable for government verification. Ideal for use in laboratories in glass apparatus.
  • Manufactured as per national and international standards
  • Enclosed scale
  • Richter top finish round
  • Available with different fillings
Fillling: Propylene carbonate ('Prop'; non-wetting by capillary coating), Paraffine Oil ('Pet'), Isoamyl benzoate ('IsoB') or hydrotreated Mineral Oil ('MinO').
Immersion depth = lower part length
* Length = upper part length
Envase: Packed in transparent square plastic case.